ADV465750068 AChemBlock

5-Iodo-2-methyl-2,3-dihydropyridazin-3-one

  • MDL number MFCD22052654
  • Cas Number : 153239-91-9
  • Molecular Weight : 236.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-iodo-2-methyl-2-3-dihydropyridazin-3-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13107
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart