ADV465750072 AChemBlock

(2R,4R)-1-[(tert-butoxy)carbonyl]-4-fluoropyrrolidine-2-carboxylic acid

  • MDL number MFCD02684479
  • Cas Number : 681128-51-8
  • Molecular Weight : 233.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-4r-1-tert-butoxy-carbonyl-4-fluoropyrrolidine-2-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13112
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart