ADV465750080 AChemBlock

6-Methoxy-1H-pyrrolo[2,3-b]pyridine-3-carboxylic acid

  • MDL number MFCD12962962
  • Cas Number : 1190317-61-3
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-methoxy-1h-pyrrolo-2-3-b-pyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13122
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart