ADV465750091 AChemBlock

4-Cyclopropyl-1H-imidazole

  • MDL number MFCD20617804
  • Cas Number : 89830-98-8
  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-cyclopropyl-1h-imidazole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13133
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart