ADV465750093 AChemBlock

3-(4-Fluorophenyl)oxetan-3-amine hydrochloride

  • MDL number MFCD19381728
  • Cas Number : 1332839-79-8
  • Molecular Weight : 203.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-4-fluorophenyl-oxetan-3-amine-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13136
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart