ADV465750100 AChemBlock

2-Chloro-7-(trifluoromethyl)quinazoline

  • MDL number MFCD22561795
  • Cas Number : 1388043-95-5
  • Molecular Weight : 232.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-chloro-7-trifluoromethyl-quinazoline.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13146
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart