ADV465750102 AChemBlock

4-Chloro-2-iodo-7-methyl-7h-pyrrolo[2,3-b]pyridine

  • MDL number MFCD29127300
  • Cas Number : 1788054-66-9
  • Molecular Weight : 292.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-2-iodo-7-methyl-7h-pyrrolo-2-3-b-pyridine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13149
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart