ADV465750108 AChemBlock

Benzyl 2,4-dichloro-5h,6h,7h,8h-pyrido[4,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD12198784
  • Cas Number : 1160248-14-5
  • Molecular Weight : 338.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/benzyl-2-4-dichloro-5h-6h-7h-8h-pyrido-4-3-d-pyrimidine-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13156
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart