ADV465750114 AChemBlock

5-Chloro-2-methylpyrazolo[1,5-a]pyrimidine

  • MDL number MFCD17676711
  • Cas Number : 189116-36-7
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-chloro-2-methylpyrazolo-1-5-a-pyrimidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13769
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart