ADV465750119 AChemBlock

1-[2-Bromo-4-(trifluoromethyl)phenyl]ethan-1-one

  • MDL number MFCD11865317
  • Cas Number : 1131605-31-6
  • Molecular Weight : 267.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-2-bromo-4-trifluoromethyl-phenyl-ethan-1-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13165
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart