ADV465750121 AChemBlock

3-Boc-8-Cbz-3,8-diazabicyclo[4.2.0]octane

  • MDL number MFCD28502575
  • Cas Number : 1630906-87-4
  • Molecular Weight : 346.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-boc-8-cbz-3-8-diazabicyclo-4-2-0-octane.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13167
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart