ADV465750125 AChemBlock

1H-Pyrrolo[2,3-b]pyridin-5-ylmethanamine

  • MDL number MFCD09475794
  • Cas Number : 267876-25-5
  • Molecular Weight : 147.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrrolo-2-3-b-pyridin-5-ylmethanamine.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13173
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart