ADV465750126 AChemBlock

5'-Amino-1',2'-dihydrospiro[cyclopropane-1,3'-indole]-2'-one

  • MDL number MFCD22420938
  • Cas Number : 1399663-06-9
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-amino-1-2-dihydrospiro-cyclopropane-1-3-indole-2-one.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13174
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart