ADV465750127 AChemBlock

Methyl 4-chloro-1-methyl-1H-pyrrolo[3,2-c]pyridine-7-carboxylate

  • MDL number MFCD17015965
  • Cas Number : 871819-43-1
  • Molecular Weight : 224.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-chloro-1-methyl-1h-pyrrolo-3-2-c-pyridine-7-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13176
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart