ADV465750136 AChemBlock

6-(Aminomethyl)-1- methyl-2-piperidinone hydrochloride

  • MDL number MFCD28501456
  • Cas Number : 1788044-00-7
  • Molecular Weight : 178.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/6-aminomethyl-1-methyl-2-piperidinone-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13186
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart