ADV465750141 AChemBlock

Methyl 2-{3-azaspiro[5.5]undecan-9-yl}acetate hydrochloride

  • MDL number MFCD28501529
  • Cas Number : 1017540-91-8
  • Molecular Weight : 261.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-3-azaspiro-5-5-undecan-9-yl-acetate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13191
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart