ADV465750142 AChemBlock

3,5-Dimethyl 6-methyl-1H-indazole-3,5-dicarboxylate

  • MDL number MFCD28501543
  • Cas Number : 1788041-62-2
  • Molecular Weight : 248.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-5-dimethyl-6-methyl-1h-indazole-3-5-dicarboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13192
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart