ADV465750144 AChemBlock

(5R)-5-(aminomethyl)-1,3-oxazolidin-2-one hydrochloride

  • MDL number MFCD28501838
  • Cas Number : 1788036-27-0
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5r-5-aminomethyl-1-3-oxazolidin-2-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13195
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart