ADV465750148 AChemBlock

tert-Butyl 1,8-diazaspiro[4.5]decane-8-carboxylate

  • MDL number MFCD11501187
  • Cas Number : 937729-06-1
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-1-8-diazaspiro-4-5-decane-8-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13199
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart