ADV465750152 AChemBlock

1-[(Benzyloxy)carbonyl]azepane-4-carboxylic acid

  • MDL number MFCD16658690
  • Cas Number : 1195256-01-9
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-benzyloxy-carbonyl-azepane-4-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13204
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart