ADV465750154 AChemBlock

5,7-Dichloro-2-methyl-2H-pyrazolo[4,3-d]pyrimidine

  • MDL number MFCD23106214
  • Cas Number : 1357087-30-9
  • Molecular Weight : 203.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-7-dichloro-2-methyl-2h-pyrazolo-4-3-d-pyrimidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13206
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart