ADV465750157 AChemBlock

tert-Butyl 2,7-diazaspiro[4.5]decane-2-carboxylate hemioxalate

  • MDL number MFCD27988097
  • Cas Number : 1523606-54-3
  • Molecular Weight : 570.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-7-diazaspiro-4-5-decane-2-carboxylate-hemioxalate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13210
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart