ADV465750167 AChemBlock

5-Chlorothieno[2,3-c]pyridine

  • MDL number MFCD18253769
  • Cas Number : 1246555-91-8
  • Molecular Weight : 169.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-chlorothieno-2-3-c-pyridine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13222
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart