ADV465750185 AChemBlock

Trans-1-Boc-3-amino-4-methylpyrrolidine

  • MDL number MFCD18375162
  • Cas Number : 1152113-30-8
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/trans-1-boc-3-amino-4-methylpyrrolidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13242
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart