ADV465750189 AChemBlock

Ethyl 1H-pyrazolo[3,4-c]pyridine-3-carboxylate

  • MDL number MFCD10566315
  • Cas Number : 1053656-33-9
  • Molecular Weight : 191.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1h-pyrazolo-3-4-c-pyridine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13247
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart