ADV465750190 AChemBlock

tert-Butyl 2-oxo-5-azaspiro[3.5]nonane-5-carboxylate

  • MDL number MFCD28501479
  • Cas Number : 1788041-51-9
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-oxo-5-azaspiro-3-5-nonane-5-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13248
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart