ADV465750191 AChemBlock

1-Acetyl-6-methyl-1H-indazole-3-carbonitrile

  • MDL number MFCD28501555
  • Cas Number : 1788043-95-7
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-acetyl-6-methyl-1h-indazole-3-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13249
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart