ADV465750198 AChemBlock

5-Bromo-6-fluoro-1H-indole-3-carbaldehyde

  • MDL number MFCD16609947
  • Cas Number : 1227496-31-2
  • Molecular Weight : 242.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-6-fluoro-1h-indole-3-carbaldehyde.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13256
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart