ADV465750199 AChemBlock

Methyl 5-chloroimidazo[1,5-a]pyridine-6-carboxylate

  • MDL number MFCD23711382
  • Cas Number : 1168090-92-3
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-5-chloroimidazo-1-5-a-pyridine-6-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13257
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart