ADV465750205 AChemBlock

Methyl 1H-pyrazolo[4,3-b]pyridine-6-carboxylate

  • MDL number MFCD20923321
  • Cas Number : 1301214-72-1
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-1h-pyrazolo-4-3-b-pyridine-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13263
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart