ADV465750212 AChemBlock

Tert-butyl 3-(bromomethyl)-3-fluoroazetidine-1-carboxylate

  • MDL number MFCD23105996
  • Cas Number : 1374658-83-9
  • Molecular Weight : 268.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3-bromomethyl-3-fluoroazetidine-1-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13272
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart