ADV465750216 AChemBlock

N-(2-{[(2r)-2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino}ethyl)morpholine-4-carboxamide hemioxalate

  • MDL number MFCD28502650
  • Cas Number : 1630815-47-2
  • Molecular Weight : 768.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/n-2-2r-2-hydroxy-3-4-hydroxyphenoxy-propyl-amino-ethyl-morpholine-4-carboxamide-hemioxalate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13277
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart