ADV465750219 AChemBlock

Cis-(3-fluoro-piperidin-4-yl)methyl-carbamic acid tert-butyl ester

  • MDL number MFCD28502736
  • Cas Number : 1376609-34-5
  • Molecular Weight : 232.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cis-3-fluoro-piperidin-4-yl-methyl-carbamic-acid-tert-butyl-ester.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13280
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart