ADV465750221 AChemBlock

Benzyl (3R,4R)-3-amino-4-hydroxypiperidine-1-carboxylate

  • MDL number MFCD22384725
  • Cas Number : 1270497-22-7
  • Molecular Weight : 250.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/benzyl-3r-4r-3-amino-4-hydroxypiperidine-1-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13282
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart