ADV465750224 AChemBlock

4-Bromo-3-methyl-1H-pyrazolo[3,4-c]pyridine

  • MDL number MFCD17016036
  • Cas Number : 1234616-30-8
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-3-methyl-1h-pyrazolo-3-4-c-pyridine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13286
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart