ADV465750230 AChemBlock

5-Methoxy-6-nitro-2,3-dihydro-1H-indole

  • MDL number MFCD23381519
  • Cas Number : 23772-38-5
  • Molecular Weight : 194.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-6-nitro-2-3-dihydro-1h-indole.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13293
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart