ADV465750231 AChemBlock

4-Amino-3,3-dimethylpyrrolidin-2-one

  • MDL number MFCD28501887
  • Cas Number : 1505180-72-2
  • Molecular Weight : 128.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-amino-3-3-dimethylpyrrolidin-2-one.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13294
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart