ADV465750240 AChemBlock

benzyl 3-((tert-butoxycarbonyl)amino)-5-hydroxypiperidine-1-carboxylate

  • MDL number MFCD26727374
  • Cas Number : 1785642-46-7
  • Molecular Weight : 350.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-piperidinecarboxylic-acid-3-1-1-dimethylethoxy-carbonyl-amino-5-hydroxy-phenylmethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13303
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart