ADV465750244 AChemBlock

4,4-Difluoro-6-azaspiro[2.5]octane hydrochloride

  • MDL number MFCD20669267
  • Cas Number : 1282532-00-6
  • Molecular Weight : 183.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-4-difluoro-6-azaspiro-2-5-octane-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13307
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart