ADV465750246 AChemBlock

Methyl (3S)-3-(2-chloropyridin-4-yl)-3-cyclopropylpropanoate

  • MDL number MFCD28501508
  • Cas Number : 1788058-39-8
  • Molecular Weight : 239.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-3s-3-2-chloropyridin-4-yl-3-cyclopropylpropanoate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13310
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart