ADV465750248 AChemBlock

methyl 2,2-dimethyl-4-oxocyclohexane-1-carboxylate

  • MDL number MFCD20926064
  • Cas Number : 1312535-32-2
  • Molecular Weight : 184.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cyclohexanecarboxylic-acid-2-2-dimethyl-4-oxo-methyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13312
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart