ADV465750254 AChemBlock

Ethyl 2-(pyridazin-3-yl)acetate

  • MDL number MFCD16036672
  • Cas Number : 1260885-52-6
  • Molecular Weight : 166.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-pyridazin-3-yl-acetate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13319
pictograms GHS05
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart