ADV465750257 AChemBlock

Methyl 2-[3-(aminomethyl)cyclobutyl]acetate hydrochloride

  • MDL number MFCD28501605
  • Cas Number : 1638765-19-1
  • Molecular Weight : 193.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-3-aminomethyl-cyclobutyl-acetate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13324
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart