ADV465750263 AChemBlock

tert-Butyl octahydro-1H-pyrrolo[3,4-c]pyridine-2-carboxylate

  • MDL number MFCD08234697
  • Cas Number : 885270-57-5
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-octahydro-1h-pyrrolo-3-4-c-pyridine-2-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13332
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart