ADV465750265 AChemBlock

(3S)-2-azabicyclo[2.2.2]octane-3-carboxylic acid

  • MDL number MFCD23105892
  • Cas Number : 109583-12-2
  • Molecular Weight : 155.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-2-azabicyclo-2-2-2-octane-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13334
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart