ADV465750266 AChemBlock

7H-Pyrrolo[2,3-c]pyridazine

  • MDL number MFCD11110436
  • Cas Number : 16767-40-1
  • Molecular Weight : 119.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/7h-pyrrolo-2-3-c-pyridazine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13335
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart