ADV465750285 AChemBlock

ABT-199

  • MDL number MFCD23160052
  • Cas Number : 1257044-40-8
  • Molecular Weight : 868.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Yellow powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/abt-199.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13360
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart