ADV465750288 AChemBlock

(1R,5S,6R)-rel-8-azabicyclo[3.2.1]octan-6-ol hydrochloride

  • MDL number MFCD28502522
  • Cas Number : 1788054-92-1
  • Molecular Weight : 163.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-5s-6r-rel-8-azabicyclo-3-2-1-octan-6-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13363
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart