ADV465750289 AChemBlock

(4S)-4-(Aminomethyl)pyrrolidin-2-one hydrochloride

  • MDL number MFCD28502527
  • Cas Number : 1638744-96-3
  • Molecular Weight : 150.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4s-4-aminomethyl-pyrrolidin-2-one-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13364
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart