ADV465750292 AChemBlock

2-(3-Chloro-4-fluorophenyl)-4,4-dimethyl-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

  • MDL number MFCD28502401
  • Cas Number : 1353971-43-3
  • Molecular Weight : 333.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-3-chloro-4-fluorophenyl-4-4-dimethyl-1-2-3-4-tetrahydroquinoline-6-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13367
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart